|
Biological Buffers
HEPES Sodium Salt
|
Printer friendly PDF version (90 Kb)
|
 |
| Sigma-Aldrich® HEPES Sodium Salt Physical Properties |
| Synonyms |
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt |
 |
| Molecular Formula |
C8H17N2O4SNa |
MDL Number |
MFCD00036463 |
Useful pH range |
6.8 - 8.2 |
| Molecular Weight |
260.29 |
EG/EC Number |
278-169-7 |
pKa (at 25) |
7.5 |
| CAS Number |
75277-39-3 |
|
| Sigma-Aldrich HEPES Sodium Salt Specifications Comparison |
| |
H3787 |
H7006 |
H3784 |
H8651 |
| Grade |
≥99.5% (titration) |
≥99.5% (titration) |
BPC, cell culture tested, ≥99.5% (titration) |
SigmaUltra, ≥99.5% (titration) |
| Purity |
≥99.5% |
≥99.5% |
≥99.5% |
≥99.5% |
| Reagent Testing |
| |
H3787 |
H7006 |
H3784 |
H8651 |
Enhanced Quality Profile Buffers |
|
X |
X |
X |
| 290NM |
|
≤0.05 |
≤0.05 |
|
| 260NM |
|
|
≤0.05 |
<0.2 |
| 280NM |
|
|
≤0.05 |
<0.2 |
| IR |
|
|
X |
|
| pH |
|
|
|
10.0 - 12.0 (20 °C, 1 M in H2O) |
| Karl Fischer |
≤7% |
≤3.0% |
≤3.0% |
|
| Insoluble Matter |
|
|
|
<0.1% |
| Total Impurities |
|
| Life Science Testing |
| |
H3787 |
H7006 |
H3784 |
H8651 |
| Endotoxin |
|
|
≤0.1 eu/mg |
|
| SPC |
|
≤20 cfu/g |
≤1000 cfu/g |
|
| Cell test |
|
|
X |
|
| RNAse |
|
|
X |
|
| DNAse |
|
|
X |
|
| Nickase |
|
|
X |
|
| Protease |
|
|
X |
|
| Trace Elemental Impurities |
| |
H3787 |
H7006 |
H3784 |
H8651 |
| Ammonia(NH4) |
|
|
|
<0.05% |
| Phosphorus(P) |
|
|
|
<0.005% |
| Chloride(Cl) |
|
|
|
<0.05% |
| Sulfate(SO4) |
|
|
|
<0.05% |
| Aluminum(Al) |
|
|
|
<0.0005% |
| Arsenic(As) |
|
| Barium(Ba) |
|
| Bismuth(Bi) |
|
| Calcium(Ca) |
|
|
|
<0.0005% |
| Cadmium(Cd) |
|
| Cobalt(Co) |
|
| Chromium(Cr) |
|
| Copper(Cu) |
|
|
|
<0.0005% |
| Iron(Fe) |
|
|
≤0.0005% |
<0.0005% |
| Potassium(K) |
|
|
|
≤0.005% |
| Lithium(Li) |
|
| Magnesium(Mg) |
|
|
|
<0.0005% |
| Manganese(Mn) |
|
| Molybdenum(Mo) |
|
| Nickel(Ni) |
|
| Lead(Pb) |
≤0.0005% |
≤0.0005% |
<0.0005% |
<0.001% |
| Strontium(Sr) |
|
| Zinc(Zn) |
|
|
|
<0.0005% |
| |
back to top
back to Biological Buffers