|
Sigma's line of coated plates for high throughput screening, epitope tag and protein capture, are available in a variety of 96- and 384-well formats. Our proprietary coating process manufactured under ISO9002 in our GMP facility delivers high well-to-well and plate-to-plate consistency creating a new standard in quality and performance!
Biotinylated Biomolecule Capture Cell-Based Assays Epitope Tag and Protein Capture Plates
For a complete specification profile of Sigma's coated multiwell plates, click here: Specification Profile
Biotinylated Biomolecule Capture
For superior binding capacity of biotinylated molecules, SigmaScreen™ Streptavidin Coated Plates offer up to 2X the binding capacity of other commercial suppliers for standard streptavidin coated plates.
| Product # |
Description |
Pack Size |
| S 8811 |
SigmaScreen Streptavidin Coated Plates, 384-well, black |
5 each 4x25 each
|
| M 4058 |
SigmaScreen Streptavidin Coated Plates, 96-well, black |
5 each 4x25 each
|
| S 8686 |
SigmaScreen Streptavidin Coated Plates, 384-well, clear |
5 each 4x25 each
|
| M 5432 |
SigmaScreen Streptavidin Coated Plates, 96-well, clear |
5 each 4x25 each
|
| S 8561 |
SigmaScreen Streptavidin Coated Plates, 384-well, white |
5 each 4x25 each
|
| M 5557 |
SigmaScreen Streptavidin Coated Plates, 96-well, white |
5 each 4x25 each
|
| M 3433 |
SigmaScreen Streptavidin Coated Strip-Well Plates, 96-well |
1 each 5 each
|
back to top
For increased sensitivity and exceptional signal-to-noise ratios, SigmaScreen™ ExtrAvidin Coated Plates are coated with a modified avidin, optimized to reduce non-specific binding.
| Product # |
Description |
Pack Size |
| E 9153 |
SigmaScreen ExtrAvidin Coated Plates, 384-well, clear |
5 each 10x5 each
|
| E 3027 |
SigmaScreen ExtrAvidin Coated Plates, 96-well, clear |
5 each 10x5 each
|
back to top
Patent pending technology allows for the highest binding capacity available today. With up to 10X more than standard streptavidin and 3X greater than other high-binding plates, Sigma's Streptavidin High Capacity Coated Plates provide a platform to capture enough peptide or protein for elution and various downstream applications.
| Product # |
Description |
Pack Size |
| S 8815 |
SigmaScreen Streptavidin High Capacity Coated Plates, 384-well, clear |
1 each 5 each
|
| S 6940 |
SigmaScreen Streptavidin High Capacity Coated Plates, 96-well, clear |
1 each 5 each
|
back to top
Cell-Based Assays
Get up to 60% greater cell adherence with SigmaScreen™ Poly-D-Lysine Coated Plates.
| Product # |
Description |
Pack Size |
| S 7686 |
SigmaScreen Poly-D-Lysine Coated Plates, 384-well, black |
5 each 5x20 each
|
| M 5307 |
SigmaScreen Poly-D-Lysine Coated Plates, 96-well, black |
5 each 5x20 each
|
| S 7436 |
SigmaScreen Poly-D-Lysine Coated Plates, 384-well, clear |
5 each 5x20 each
|
| M 3555 |
SigmaScreen Poly-D-Lysine Coated Plates, 96-well, clear |
5 each 5x20 each
|
| S 7561 |
SigmaScreen Poly-D-Lysine Coated Plates, 384-well, white |
5 each 5x20 each
|
| M 5682 |
SigmaScreen Poly-D-Lysine Coated Plates, 96-well, white |
5 each 5x20 each
|
All PDL plate formats are clear bottom and include lids.
back to top
SigmaScreen™ Collagen I Coated Plates offer rapid and uniform cell proliferation.
| Product # |
Description |
Pack Size |
| S 3690 |
SigmaScreen Collagen I Coated Plates, 384-well, black |
5 each 10x5 each
|
| S 3190 |
SigmaScreen Collagen I Coated Plates, 96-well, black |
5 each 10x5 each
|
| S 3440 |
SigmaScreen Collagen I Coated Plates, 384-well, clear |
5 each 10x5 each
|
| S 8813 |
SigmaScreen Collagen I Coated Plates, 96-well, clear |
5 each 10x5 each
|
| S 3565 |
SigmaScreen Collagen I Coated Plates, 384-well, white |
5 each 10x5 each
|
| S 3315 |
SigmaScreen Collagen I Coated Plates, 96-well, white |
5 each 10x5 each
|
All Collagen I plate formats are clear bottom and include lids.
back to top
SigmaScreen™ Fibronectin Coated Plates provide a superior environment for promoting cell-plating efficiency and improving morphology.
| Product # |
Description |
Pack Size |
| S 3815 |
SigmaScreen™ Fibronectin Coated Multiwell Plate, 96 well clear |
5 each 10 x 5 each
|
back to top
Epitope Tag and Protein Capture Plates
| Product # |
Description |
Pack Size |
| P 2241 |
Anti c-Myc Coated 96-well, clear |
1 each 5 each
|
| S 5563 |
HIS-Select™ HC Nickel Coated 96-well, clear |
1 each 5 each
|
| S 5688 |
HIS-Select™ HS Nickel Coated 96-well, strip |
1 each 5 each
|
| H 9412 |
HIS-Select™ iLAP™ High Capacity (HC) Nickel Coated 96-well, clear |
1 each 5 each
|
| P 2983 |
Anti-FLAG™ HS M2 Coated 96-well, clear |
1 each 5 each
|
| G3294 |
Glutathione High Capacity (HC) Coated 96-well, clear |
1 each 5 each
|
| P 3233 |
Glutathione HS Coated 96-well, clear |
1 each 5 each
|
| P1617 |
PHOS-Select High Capacity (HC) Iron Coated 96-well, clear |
1 each 5 each
|
| S1938 |
Protein A High Sensitivity (HS) Coated 96-well, clear |
1 each 5 each
|
| S 2063 |
Protein G HS Coated 96-well, clear |
1 each 5 each
|
back to top
Sigma's line of coated plates utilize Corning® base plates.
To Order Call: 1-800-325-3010 Technical Service: 1-800-325-5832
Corning is a registered trademark of Corning Incorporated, Corning, NY 14831-0001. SigmaScreen is a trademark of Sigma-Aldrich Corporation, St. Louis, MO 63103.
|